1-(2,3-Difluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65979-500MG 65979-1G 65979-5G 65979-25G
77332-692EA 258.53 USD
77332-692 77356-948 77368-738 77389-924
1-(2,3-Difluorophenyl)-2,2-difluoroethanone ≥97%
1-(2,3-Difluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MW: 192.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045199
CAS Number: 1386985-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR