2,2-Difluoro-1-(3-fluoro-5-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65976-500MG 65976-1G 65976-5G 65976-25G
77332-688EA 184.37 USD
77332-688 77347-290 77368-736 77389-922
2,2-Difluoro-1-(3-fluoro-5-methylphenyl)ethanone ≥97%
2,2-Difluoro-1-(3-fluoro-5-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MW: 188.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920996
CAS Number: 1352215-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR