1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65975-500MG 65975-1G 65975-5G 65975-25G
77332-686EA 184.37 USD
77332-686 77347-288 77368-734 77389-920
1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2-difluoroethanone ≥97%
1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD34762345
CAS Number: 2807467-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR