1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65972-500MG 65972-1G 65972-5G 65972-25G
77332-682EA 257.5 USD
77332-682 77356-940 77368-730 77389-916
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34762344
CAS Number: 2807446-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR