1-(6-Bromo-3-chloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65970-500MG 65970-1G 65970-5G 65970-25G
77332-678EA 182.31 USD
77332-678 77347-002 77368-726 77389-912
1-(6-Bromo-3-chloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(6-Bromo-3-chloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃BrClF₃O
MW: 287.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762343
CAS Number: 2229074-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR