2,2-Difluoro-1-(5-fluoro-2-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65965-500MG 65965-1G 65965-5G 65965-25G
77332-672EA 202.91 USD
77332-672 77356-938 77368-720 77389-908
2,2-Difluoro-1-(5-fluoro-2-(trifluoromethyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(5-fluoro-2-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄F₆O
MW: 242.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31807463
CAS Number: 2091613-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR