1-(2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
65964-500MG
65964-1G
65964-5G
65964-25G
77332-670EA
260.59
USD
77332-670
77356-936
77368-718
77389-906
1-(2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥97%
1-(2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₃BrF₆O MW: 321.014 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762341 CAS Number: 2807441-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...