2,2-Difluoro-1-(3-fluoro-4-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65963-500MG 65963-1G 65963-5G 65963-25G
77332-668EA 184.37 USD
77332-668 77347-286 77368-716 77389-904
2,2-Difluoro-1-(3-fluoro-4-methylphenyl)ethanone ≥97%
2,2-Difluoro-1-(3-fluoro-4-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MW: 188.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920987
CAS Number: 1352217-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR