1-(6-Bromo-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65962-500MG 65962-1G 65962-5G 65962-25G
77332-666EA 161.71 USD
77332-666 77345-954 77368-714 77389-902
1-(6-Bromo-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥97%
1-(6-Bromo-2-fluoro-3-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O
MW: 267.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762340
CAS Number: 2807467-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR