1-(3,5-Difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65959-500MG 65959-1G 65959-5G 65959-25G
77332-662EA 178.19 USD
77332-662 77346-678 77368-712 77389-898
1-(3,5-Difluorophenyl)-2,2-difluoroethanone ≥95%
1-(3,5-Difluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MW: 192.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920690
CAS Number: 1352205-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR