1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65955-500MG 65955-1G 65955-5G 65955-25G
77332-658EA 141.11 USD
77332-658 77344-868 77368-708 77389-894
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
MW: 283.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762337
CAS Number: 2807444-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR