2,2-Difluoro-1-(3-(trifluoromethoxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65954-500MG 65954-1G 65954-5G 65954-25G
77332-656EA 178.19 USD
77332-656 77346-676 77368-706 77389-892
2,2-Difluoro-1-(3-(trifluoromethoxy)phenyl)ethanone ≥95%
2,2-Difluoro-1-(3-(trifluoromethoxy)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O₂
MW: 240.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045151
CAS Number: 1566866-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR