1-(4-Ethoxy-3-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65953-500MG 65953-1G 65953-5G 65953-25G
77332-654EA 82.4 USD
77332-654 77342-180 77368-704 77380-886
1-(4-Ethoxy-3-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(4-Ethoxy-3-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762336
CAS Number: 2901101-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR