1-(3,6-Dibromo-2-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65949-500MG 65949-1G 65949-5G 65949-25G
77332-652EA 233.81 USD
77332-652 77356-930 77368-700 77389-890
1-(3,6-Dibromo-2-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(3,6-Dibromo-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃Br₂F₃O
MW: 331.912 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762334
CAS Number: 2807474-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR