1-(2-Bromo-6-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65947-500MG 65947-1G 65947-5G 65947-25G
77332-650EA 202.91 USD
77332-650 77356-928 77368-698 77389-888
1-(2-Bromo-6-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(2-Bromo-6-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MW: 253.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762333
CAS Number: 2090411-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR