1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65946-500MG 65946-1G 65946-5G 65946-25G
77332-648EA 157.59 USD
77332-648 77345-708 77368-696 77389-886
1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃BrClF₃O
MW: 287.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29914187
CAS Number: 2090671-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR