3-Bromo-2-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65945-500MG 65945-1G 65945-5G 65945-25G
77332-646EA 271.92 USD
77332-646 77356-926 77368-694 77389-884
3-Bromo-2-fluoro-1,1'-biphenyl ≥97%
3-Bromo-2-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD26687708
CAS Number: 1515573-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR