1-(3-Bromo-2,6-difluoro-5-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65938-500MG 65938-1G 65938-5G
77338-364EA 481.01 USD
77338-364 77356-924 77380-882
1-(3-Bromo-2,6-difluoro-5-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Bromo-2,6-difluoro-5-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₄BrF₅O₂
Storage Temperature: Ambient
MDL Number: MFCD34762330
CAS Number: 2807450-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR