4'-Bromo-2'-ethyl-2,3-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65917-500MG 65917-1G 65917-5G 65917-25G
77332-644EA 161.71 USD
77332-644 77345-952 77368-692 77389-882
4'-Bromo-2'-ethyl-2,3-dimethyl-1,1'-biphenyl ≥95%
4'-Bromo-2'-ethyl-2,3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762325
CAS Number: 2807470-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR