2-Chloro-4-fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65908-500MG 65908-1G 65908-5G 65908-25G
77332-634EA 157.59 USD
77332-634 77345-660 77368-684 77389-874
2-Chloro-4-fluoro-3-iodobenzaldehyde ≥97%
2-Chloro-4-fluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
MW: 284.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762321
CAS Number: 2383782-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR