3-Bromo-2-chloro-4-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

65906-500MG 65906-1G 65906-5G 65906-25G
77332-632EA 204.97 USD
77332-632 77356-920 77368-682 77389-872
3-Bromo-2-chloro-4-fluorobenzaldehyde ≥95%
3-Bromo-2-chloro-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
Storage Temperature: Ambient
MDL Number: MFCD26688000
CAS Number: 1536695-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR