1-(5-Chloro-2-isopropoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65845-500MG 65845-1G 65845-5G
77332-622EA 338.87 USD
77332-622 77356-912 77380-874
1-(5-Chloro-2-isopropoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(5-Chloro-2-isopropoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34762316
CAS Number: 1552104-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR