1-(3,6-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanol ≥97%
Supplier: AOBChem USA
|
Warning
|
65830-500MG
65830-1G
65830-5G
65830-25G
77332-618EA
242.05
USD
77332-618
77356-908
77368-670
77389-862
1-(3,6-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanol ≥97%
1-(3,6-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanol
Precaución: for Research Use Only
Formula:
C₈H₄Cl₂F₄O MW: 263.016 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762312 CAS Number: 2901098-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...