1-Chloro-4-iodo-3-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

65812-500MG 65812-1G 65812-5G
77340-348EA 912.58 USD
77340-348 77356-906 77380-872
1-Chloro-4-iodo-3-methoxy-2-methylbenzene ≥97%
1-Chloro-4-iodo-3-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
Storage Temperature: Ambient
MDL Number: MFCD34762306
CAS Number: 2386989-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR