(4-Ethoxy-2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

65747-500MG 65747-1G 65747-5G 65747-25G
77332-610EA 211.15 USD
77332-610 77356-900 77368-662 77389-856
(4-Ethoxy-2-fluorophenyl)methanol ≥97%
(4-Ethoxy-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD28519144
CAS Number: 1393470-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR