1-(3,5-Dimethyl-4-propoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65719-500MG 65719-1G 65719-5G 65719-25G
77332-608EA 243.08 USD
77332-608 77356-898 77368-660 77389-854
1-(3,5-Dimethyl-4-propoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,5-Dimethyl-4-propoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD16660672
CAS Number: 1443324-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR