2,2,2-Trifluoro-1-(5-fluoro-2-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65703-500MG 65703-1G 65703-5G 65703-25G
77332-606EA 215.27 USD
77332-606 77356-896 77368-658 77389-852
2,2,2-Trifluoro-1-(5-fluoro-2-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(5-fluoro-2-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₃F₇O
MW: 260.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31807462
CAS Number: 2228702-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR