2,2,2-Trifluoro-1-(2-fluoro-4-(trifluoromethyl)phenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
65698-500MG
65698-1G
65698-5G
65698-25G
77332-602EA
264.71
USD
77332-602
77356-894
77368-654
77389-850
2,2,2-Trifluoro-1-(2-fluoro-4-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-4-(trifluoromethyl)phenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₃F₇O MW: 260.108 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30481122 CAS Number: 1252780-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...