2,2,2-Trifluoro-1-(2-(trifluoromethoxy)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65684-500MG 65684-1G 65684-5G 65684-25G
77332-600EA 122.57 USD
77332-600 77344-004 77368-652 77380-864
2,2,2-Trifluoro-1-(2-(trifluoromethoxy)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-(trifluoromethoxy)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄F₆O₂
MW: 258.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18046777
CAS Number: 1281667-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR