2-(3-Fluoro-2,4-bis(methylthio)phenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

65681-500MG 65681-1G 65681-5G 65681-25G
77332-598EA 256.47 USD
77332-598 77356-892 77368-650 77389-848
2-(3-Fluoro-2,4-bis(methylthio)phenyl)-1,3-dioxolane ≥95%
2-(3-Fluoro-2,4-bis(methylthio)phenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂S₂
MW: 260.348 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762285
CAS Number: 2807448-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR