2,2,2-Trifluoro-1-(2-hydroxy-5-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65678-500MG 65678-1G 65678-5G
77336-546EA 388.31 USD
77336-546 77356-890 77380-862
2,2,2-Trifluoro-1-(2-hydroxy-5-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-hydroxy-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄F₆O₂
MW: 258.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762284
CAS Number: 2102203-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR