2,2,2-Trifluoro-1-(4-(trifluoromethoxy)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65676-500MG 65676-1G 65676-5G 65676-25G
77332-596EA 153.47 USD
77332-596 77345-518 77368-648 77389-846
2,2,2-Trifluoro-1-(4-(trifluoromethoxy)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-(trifluoromethoxy)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄F₆O₂
MW: 258.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370307
CAS Number: 107713-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR