2,2,2-Trifluoro-1-(4-fluoronaphthalen-1-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65660-500MG 65660-1G 65660-5G 65660-25G
77332-594EA 126.69 USD
77332-594 77344-192 77368-644 77380-856
2,2,2-Trifluoro-1-(4-fluoronaphthalen-1-yl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-fluoronaphthalen-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₆F₄O
MW: 242.169 g/mol
Storage Temperature: Ambient
CAS Number: 664364-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR