6-Chloro-7-fluoro-3-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

65603-500MG 65603-1G 65603-5G
77336-652EA 390.37 USD
77336-652 77356-886 77380-850
6-Chloro-7-fluoro-3-methyl-1H-indazole ≥97%
6-Chloro-7-fluoro-3-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆ClFN₂
MW: 184.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762278
CAS Number: 2807450-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR