1-(3-Chloro-4-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65590-500MG 65590-1G 65590-5G 65590-25G
77332-588EA 188.49 USD
77332-588 77356-880 77368-636 77389-842
1-(3-Chloro-4-methylphenyl)-2,2-difluoroethanone ≥95%
1-(3-Chloro-4-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O
MW: 204.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920499
CAS Number: 1352222-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR