1-(3,5-Dichlorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65582-500MG 65582-1G 65582-5G 65582-25G
77332-586EA 178.19 USD
77332-586 77346-618 77368-634 77389-840
1-(3,5-Dichlorophenyl)-2,2-difluoroethanone ≥97%
1-(3,5-Dichlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄Cl₂F₂O
MW: 225.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618112
CAS Number: 1253787-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR