1-(4-Chloro-2-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65572-500MG 65572-1G 65572-5G
77336-928EA 400.67 USD
77336-928 77356-878 77380-844
1-(4-Chloro-2-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25981802
CAS Number: 1566995-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR