1-(6-Chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65566-500MG 65566-1G 65566-5G 65566-25G
77332-582EA 141.11 USD
77332-582 77344-866 77368-632 77389-838
1-(6-Chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
1-(6-Chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762265
CAS Number: 2807467-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR