(6-Chloro-2-fluoro-3-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

65564-500MG 65564-1G 65564-5G 65564-25G
77332-580EA 120.51 USD
77332-580 77343-790 77368-630 77380-840
(6-Chloro-2-fluoro-3-methylphenyl)trimethylsilane ≥97%
(6-Chloro-2-fluoro-3-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClFSi
MW: 216.755 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762263
CAS Number: 2807441-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR