2-Bromo-1-chloro-3-ethoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

65558-500MG 65558-1G 65558-5G
77337-812EA 445.99 USD
77337-812 77356-874 77380-838
2-Bromo-1-chloro-3-ethoxy-4-methylbenzene ≥97%
2-Bromo-1-chloro-3-ethoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762259
CAS Number: 2384606-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR