2-Bromo-1-chloro-3-methoxy-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

65550-500MG 65550-1G 65550-5G
77338-432EA 487.19 USD
77338-432 77356-872 77380-836
2-Bromo-1-chloro-3-methoxy-4-methylbenzene ≥95%
2-Bromo-1-chloro-3-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33398429
CAS Number: 1783711-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR