1-(Benzyloxy)-2,4-difluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

65539-500MG 65539-1G 65539-5G 65539-25G
77332-578EA 264.71 USD
77332-578 77356-870 77368-628 77389-836
1-(Benzyloxy)-2,4-difluoro-5-iodobenzene ≥97%
1-(Benzyloxy)-2,4-difluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉F₂IO
MW: 346.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762253
CAS Number: 2807450-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR