1-(4-Chloro-3,5-dimethylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65535-500MG 65535-1G 65535-5G 65535-25G
77332-576EA 260.59 USD
77332-576 77356-868 77368-626 77389-834
1-(4-Chloro-3,5-dimethylphenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-3,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₂O
MW: 218.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103710
CAS Number: 1823319-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR