4,4'-Dibromo-2,6-difluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65527-500MG 65527-1G 65527-5G 65527-25G
77332-574EA 184.37 USD
77332-574 77347-276 77368-622 77389-832
4,4'-Dibromo-2,6-difluoro-1,1'-biphenyl ≥95%
4,4'-Dibromo-2,6-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆Br₂F₂
Storage Temperature: Ambient
MDL Number: MFCD34762246
CAS Number: 2807441-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR