1-(Benzyloxy)-5-chloro-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

65519-500MG 65519-1G 65519-5G
77332-572EA 329.6 USD
77332-572 77356-866 77380-832
1-(Benzyloxy)-5-chloro-2-fluoro-3-methylbenzene ≥97%
1-(Benzyloxy)-5-chloro-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO
MW: 250.696 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762239
CAS Number: 2901098-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR