1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65509-500MG 65509-1G 65509-5G 65509-25G
77332-568EA 264.71 USD
77332-568 77356-860 77368-618 77389-828
1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅BrF₄OS
MW: 317.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762235
CAS Number: 2807464-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR