4'-Bromo-4-chloro-3-isopropyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65508-500MG 65508-1G 65508-5G
77337-210EA 408.91 USD
77337-210 77356-858 77380-828
4'-Bromo-4-chloro-3-isopropyl-1,1'-biphenyl ≥97%
4'-Bromo-4-chloro-3-isopropyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrCl
MW: 309.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762234
CAS Number: 2807471-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR