4-Chloro-3-isopropyl-4'-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65507-500MG 65507-1G 65507-5G
77337-986EA 456.29 USD
77337-986 77356-856 77380-826
4-Chloro-3-isopropyl-4'-methoxy-1,1'-biphenyl ≥97%
4-Chloro-3-isopropyl-4'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇ClO
MW: 260.759 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762233
CAS Number: 2807445-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR