4-Chloro-4'-fluoro-3-isopropyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65506-500MG 65506-1G 65506-5G
77335-718EA 363.59 USD
77335-718 77356-854 77380-824
4-Chloro-4'-fluoro-3-isopropyl-1,1'-biphenyl ≥97%
4-Chloro-4'-fluoro-3-isopropyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₄ClF
MW: 248.723 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762232
CAS Number: 2807455-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR