1-(6-Chloro-2-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
65496-500MG
65496-1G
65496-5G
65496-25G
77332-560EA
178.19
USD
77332-560
77346-672
77368-610
77389-822
1-(6-Chloro-2-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(6-Chloro-2-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₅ClF₄O MW: 240.582 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26680899 CAS Number: 1538384-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...